Learn More
AdipoGen Pyrithiobac
SDP
Supplier:  AdipoGen CDXP0131M250
Encompass_Preferred
Chemical. CAS 123342-93-8. Formula C13H11ClN2O4S. MW 326.8. Synthetic. Aromatic acid herbicide. Mode of action is through inhibition of acetolactate synthase ALS acetohydroxyacid synthase AHAS. Compound can be used as analytical reference material.
Catalog No. 50-256-0232