Learn More
Medchemexpress LLC Pyrithione | 1121-30-8 | 99.8% | 50 MG
SDP
Catalog No. 5000412243
Encompass_Preferred
Pyrithione is an inhibitor of membrane transport processes in fungi and a potent antibiotic aspergillic acid. It significantly decreases the activities of various independently regulated transport systems when Penicillium mycelia are incubated with the compound. It inhibits 35SO42- transport and L-[14C]-phenylalanine transport, and reduces ATP levels in Penicillium chrysogenum.
  • Inhibits membrane transport processes in fungi
  • Potent antibiotic aspergillic acid
  • Decreases activities of various transport systems
  • Reduces ATP level
  • Prevents incorporation of accumulated L-[14C]tryptophan and 35SO42- into protein
Catalog No. 50-004-12243 Supplier Medchemexpress LLC Supplier No. HYB174750MG
Pyrithione is an inhibitor of membrane transport processes in fungi and a potent antibiotic aspergillic acid. It significantly decreases the activities of various independently regulated transport systems when Penicillium mycelia are incubated with the compound. It inhibits 35SO42- transport and L-[14C]-phenylalanine transport, and reduces ATP levels in Penicillium chrysogenum.
  • Inhibits membrane transport processes in fungi
  • Potent antibiotic aspergillic acid
  • Decreases activities of various transport systems
  • Reduces ATP level
  • Prevents incorporation of accumulated L-[14C]tryptophan and 35SO42- into protein