Learn More
Medchemexpress LLC Pyrithione | 1121-30-8 | MFCD00006196 | 99.8% | 5 MG
SDP
Catalog No. 5000412130
Encompass_Preferred
Pyrithione is an inhibitor of membrane transport processes in fungi and acts as a potent antibiotic aspergillic acid. When Penicillium mycelia are incubated with Pyrithione, a significant decrease in the activities of various independently regulated transport systems is observed.
  • Inhibits membrane transport processes in fungi.
  • Functions as a potent antibiotic aspergillic acid.
  • Decreases activities of various independently regulated transport systems.
  • Inhibits 35SO42- transport in P. notatum in vitro (IC50 = 0.6 mM).
  • Inhibits L-[14C]-phenylalanine transport in P. chrysogenum in vitro (IC50 = 0.6 mM).
  • Reduces ATP levels in P. chrysogenum to about 40% of initial level at 2.5 mM.
  • Prevents incorporation of internal accumulated L-[14C]tryptophan and 35SO42- into protein at 5 mM.
Catalog No. 50-004-12130 Supplier Medchemexpress LLC Supplier No. HYB17475MG
Pyrithione is an inhibitor of membrane transport processes in fungi and acts as a potent antibiotic aspergillic acid. When Penicillium mycelia are incubated with Pyrithione, a significant decrease in the activities of various independently regulated transport systems is observed.
  • Inhibits membrane transport processes in fungi.
  • Functions as a potent antibiotic aspergillic acid.
  • Decreases activities of various independently regulated transport systems.
  • Inhibits 35SO42- transport in P. notatum in vitro (IC50 = 0.6 mM).
  • Inhibits L-[14C]-phenylalanine transport in P. chrysogenum in vitro (IC50 = 0.6 mM).
  • Reduces ATP levels in P. chrysogenum to about 40% of initial level at 2.5 mM.
  • Prevents incorporation of internal accumulated L-[14C]tryptophan and 35SO42- into protein at 5 mM.