Learn More
TARGETMOL CHEMICALS INC Pyrithione 50MG
SDP
Catalog No. 502879572
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. Pyrithione (1-hydroxyl-1H-pyridine-2-thione) is a transition metal complex, a zinc ionophore, that causes elevated zinc levels in mammalian cells. Pyrithione has potent bactericidal and antifungal activity.
Catalog No. 50-287-9572 Supplier TARGETMOL CHEMICALS INC Supplier No. T1259350MG
Also available in 5 mg, 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. Pyrithione (1-hydroxyl-1H-pyridine-2-thione) is a transition metal complex, a zinc ionophore, that causes elevated zinc levels in mammalian cells. Pyrithione has potent bactericidal and antifungal activity.