Learn More
Selleck Chemical LLC Pyrithione-E1774-5MG
SDP
Supplier:  Selleck Chemical LLC E17745MG
Encompass_Preferred
Pyrithione a Transition metal complexe is a zinc ionophore that causes increased zinc levels within mammalian cells Pyrithione has potent bactericidal and anti-fungal activity
Catalog No. 50-313-8890