Learn More
Medchemexpress LLC Pyrithioxin dihydrochloride | 10049-83-9 | 98.0% | 10 MM 1 ML
SDP
Catalog No. 5000408473
Encompass_Preferred
Pyrithioxin (Pyritinol) dihydrochloride is the dihydrochloride salt of Pyrithioxin, an orally active neurodynamic compound. It promotes the metabolism of glucose and amino acids, increases carotid blood flow, and improves cerebral blood flow. Pyrithioxin exhibits anti-inflammation, anti-tumor, and neuroprotective effects. It is used in research related to cancer, inflammation, immunology, metabolic, and neurological diseases such as cerebral infarct, epilepsy, fibrosarcomas, and rheumatoid polyarthritis.
  • Orally active neurodynamic compound
  • Promotes glucose and amino acid metabolism
  • Increases carotid blood flow
  • Improves cerebral blood flow
  • Exhibits anti-inflammation effect
  • Exhibits anti-tumor effect
  • Exhibits neuroprotective effect
  • Used for research in cancer, inflammation, immunology, metabolic, and neurological diseases
Catalog No. 50-004-08473 Supplier Medchemexpress LLC Supplier No. HYB0910A10MM/1ML
Pyrithioxin (Pyritinol) dihydrochloride is the dihydrochloride salt of Pyrithioxin, an orally active neurodynamic compound. It promotes the metabolism of glucose and amino acids, increases carotid blood flow, and improves cerebral blood flow. Pyrithioxin exhibits anti-inflammation, anti-tumor, and neuroprotective effects. It is used in research related to cancer, inflammation, immunology, metabolic, and neurological diseases such as cerebral infarct, epilepsy, fibrosarcomas, and rheumatoid polyarthritis.
  • Orally active neurodynamic compound
  • Promotes glucose and amino acid metabolism
  • Increases carotid blood flow
  • Improves cerebral blood flow
  • Exhibits anti-inflammation effect
  • Exhibits anti-tumor effect
  • Exhibits neuroprotective effect
  • Used for research in cancer, inflammation, immunology, metabolic, and neurological diseases