Learn More
Medchemexpress LLC Pyrithioxin dihydrochloride | 10049-83-9 | 98.0% | 500 MG
SDP
Catalog No. 5000408661
Encompass_Preferred
Pyrithioxin (Pyritinol) dihydrochloride is the dihydrochloride salt of Pyrithioxin. Pyrithioxin (Pyritinol) is an orally active neurodynamic compound. Pyrithioxin can promote the metabolism of glucose and amino acids, increase carotid blood flow and improve cerebral blood flow. Pyrithioxin exhibits anti-inflammation, anti-tumor and neuroprotective effect. Pyrithioxin can be used for the researches of cancer, inflammation, immunology, metabolic and neurological disease such as cerebral infarct, epilepsy, fibrosarcomas and rheumatoid polyarthritis.
  • Promotes metabolism of glucose and amino acids
  • Increases carotid blood flow and improves cerebral blood flow
  • Exhibits anti-inflammation effects
  • Exhibits anti-tumor effects
  • Provides neuroprotective effects
Catalog No. 50-004-08661 Supplier Medchemexpress LLC Supplier No. HYB0910A500MG
Pyrithioxin (Pyritinol) dihydrochloride is the dihydrochloride salt of Pyrithioxin. Pyrithioxin (Pyritinol) is an orally active neurodynamic compound. Pyrithioxin can promote the metabolism of glucose and amino acids, increase carotid blood flow and improve cerebral blood flow. Pyrithioxin exhibits anti-inflammation, anti-tumor and neuroprotective effect. Pyrithioxin can be used for the researches of cancer, inflammation, immunology, metabolic and neurological disease such as cerebral infarct, epilepsy, fibrosarcomas and rheumatoid polyarthritis.
  • Promotes metabolism of glucose and amino acids
  • Increases carotid blood flow and improves cerebral blood flow
  • Exhibits anti-inflammation effects
  • Exhibits anti-tumor effects
  • Provides neuroprotective effects