Learn More
Cayman Chemical Pyrantel (pamoate), Purity: ≥95%, CAS: 22204-24-6, 50 g
SDP
Catalog No. 502940069
Encompass_Preferred
An anthelmintic and agonist of nAChRs; activates chimeric C. elegans α7-nAChR expressed in BOSC cells (EC50 = 40 μM); induces contraction of A. suum muscle (EC50 = 57.5 nM); decreases survival of A. ceylanicum, N. americanus, and T. muris third-stage larvae (IC50s = 90.9, 2, and 95.5 μg/ml, respectively) and N. americanus and T. muris adults (IC50s = 7.6 and 34.1 μg/ml, respectively) in vitro; decreases intestinal A. ceylanicum burden in hamsters and intestinal H. bakeri burden in mice at 6 mg/kg
Catalog No. 50-294-0069 Supplier Cayman Chemical Supplier No. 2736850 G
An anthelmintic and agonist of nAChRs; activates chimeric C. elegans α7-nAChR expressed in BOSC cells (EC50 = 40 μM); induces contraction of A. suum muscle (EC50 = 57.5 nM); decreases survival of A. ceylanicum, N. americanus, and T. muris third-stage larvae (IC50s = 90.9, 2, and 95.5 μg/ml, respectively) and N. americanus and T. muris adults (IC50s = 7.6 and 34.1 μg/ml, respectively) in vitro; decreases intestinal A. ceylanicum burden in hamsters and intestinal H. bakeri burden in mice at 6 mg/kg