Learn More
Perkin Elmer US LLC HGA Graphite Walled Tubes, Pkg. 10
Catalog No. 509051965
Encompass_Preferred
Standard pyrocoated graphite walled tubes are produced in a quality controlled manner to ensure reliable temperature calibration in PerkinElmer graphite furnaces.
Catalog No. 50-905-1965 Supplier Perkin Elmer US LLC Supplier No. B0135653
Standard pyrocoated graphite walled tubes are produced in a quality controlled manner to ensure reliable temperature calibration in PerkinElmer graphite furnaces.