Learn More
Lonza Walkersville Pyrogen-free Test Tubes
Catalog No. NC9489157
Encompass
UPDATE (Apr 2019):We have decided to discontinue the performance of a sterility test for our Pyrogen-free Tubes.As part of the production process, all glassware is depyrogenated at extremely high temperatures well in excess of standard depyrogenation process conditions. These conditions provide for ample assurance that the tubes are depyrogenated making them suitable for use when performing the Bacterial Endotoxins Test (BET). Therefore, there is no added value or additional quality assurance in subsequently performing a sterility test after processing. As a result, the Certificate of Analyses will no longer indicate the results of a sterility test for these products.The implementation date for this change is the end of April 2019.Benefits:Certified to contain less than 0.005 EU/ml endotoxin
Catalog No. NC9489157 Supplier Lonza Walkersville Supplier No. N205
UPDATE (Apr 2019):We have decided to discontinue the performance of a sterility test for our Pyrogen-free Tubes.As part of the production process, all glassware is depyrogenated at extremely high temperatures well in excess of standard depyrogenation process conditions. These conditions provide for ample assurance that the tubes are depyrogenated making them suitable for use when performing the Bacterial Endotoxins Test (BET). Therefore, there is no added value or additional quality assurance in subsequently performing a sterility test after processing. As a result, the Certificate of Analyses will no longer indicate the results of a sterility test for these products.The implementation date for this change is the end of April 2019.Benefits:Certified to contain less than 0.005 EU/ml endotoxin