Learn More
Sigma Aldrich Fine Chemicals Biosciences Pyrocatechol Violet suitab10G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P788410G
Encompass
Pyrocatechol Violet (PV) is a sulfone phthalein dye. It chelates metal ions. The metal-PV complex is bluish-violet in color.
Catalog No. 50-289-7441