Learn More
Medchemexpress LLC HY-Y0202 100mg Medchemexpress, Pyrocatechuic acid CAS:303-38-8 Purity:>98%
SDP
Catalog No. 501970756
Encompass_Preferred
Medchemexpress, HY-Y0202 100mg Pyrocatechuic acid CAS:303-38-8 Pyrocatechuic acid is a normal human benzoic acid metabolite found in plasma, and has increased levels after aspirin ingestion. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0756 Supplier Medchemexpress LLC Supplier No. HYY0202100MG
Medchemexpress, HY-Y0202 100mg Pyrocatechuic acid CAS:303-38-8 Pyrocatechuic acid is a normal human benzoic acid metabolite found in plasma, and has increased levels after aspirin ingestion. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.