Learn More
Lonza Walkersville Lonza PyroCell™ Monocyte Activation Test Kit, In Vitro, Pyrogen Testing
Catalog No. NC3986794
Encompass
The PyroCell™ Monocyte Activation Test (MAT) Kit is an in vitro assay for pyrogen testing in biopharmaceutical and medical device industries. It detects inflammatory cytokines released by human monocytes in response to endotoxins and non-endotoxin pyrogens, aligning with European Pharmacopeia and 3Rs principles.
  • Sustainable pyrogen test system, avoiding animal use.
  • Regulatory compliant; detects full range of pyrogens.
  • Qualified cells ensure reliable, reproducible results.
  • Optimized assay components for high sensitivity.
  • Fully compatible with hardware.
  • Tests raw materials, pharmaceuticals, medical devices.
  • Supports in-process pyrogen testing.
  • Ideal for biological products (proteins, antibodies, vaccines).
Catalog No. NC3986794 Supplier Lonza Walkersville Supplier No. 00249735
The PyroCell™ Monocyte Activation Test (MAT) Kit is an in vitro assay for pyrogen testing in biopharmaceutical and medical device industries. It detects inflammatory cytokines released by human monocytes in response to endotoxins and non-endotoxin pyrogens, aligning with European Pharmacopeia and 3Rs principles.
  • Sustainable pyrogen test system, avoiding animal use.
  • Regulatory compliant; detects full range of pyrogens.
  • Qualified cells ensure reliable, reproducible results.
  • Optimized assay components for high sensitivity.
  • Fully compatible with hardware.
  • Tests raw materials, pharmaceuticals, medical devices.
  • Supports in-process pyrogen testing.
  • Ideal for biological products (proteins, antibodies, vaccines).