Learn More
Associates of Cape Cod Inc PYROCLEAR 10 X 75MM REACTION
SDP
Catalog No. NC0853843
Encompass
Pyrotubes. 10 x 75 mm soda lime glass for gel-clot method, 50 tubes/pack, 10 packs/carton
Catalog No. NC0853843 Supplier Associates of Cape Cod Inc Supplier No. TS05010
Pyrotubes. 10 x 75 mm soda lime glass for gel-clot method, 50 tubes/pack, 10 packs/carton