Learn More
Associates of Cape Cod Inc PYROCLEAR PIPETTE TIPS
SDP
Catalog No. NC0938050
Encompass
Precision Pipette Tips, 250 �l Tips, 96 Tips/Box, 10 Boxes/Pack Our Pyroclear� Precision Pipette Tips Are Certified Free Of Interfering Endotoxins And Glucans.
Catalog No. NC0938050 Supplier Associates of Cape Cod Inc Supplier No. PPT25
Precision Pipette Tips, 250 �l Tips, 96 Tips/Box, 10 Boxes/Pack Our Pyroclear� Precision Pipette Tips Are Certified Free Of Interfering Endotoxins And Glucans.