Learn More
Perkin Elmer US LLC Integrated Platform HGA Graphite Tubes, Pkg. 20
Catalog No. 509052076
Encompass_Preferred
Graphite tubes with integrated platforms are machined from one piece of graphite and are totally pyrocoated, providing exceptional tube-to-tube reproducibility.
Catalog No. 50-905-2076 Supplier Perkin Elmer US LLC Supplier No. B3001264
Graphite tubes with integrated platforms are machined from one piece of graphite and are totally pyrocoated, providing exceptional tube-to-tube reproducibility.