Learn More
Medchemexpress LLC HY-N1579 1g Medchemexpress, Pyrogallol CAS:87-66-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN15791G
Encompass_Preferred
Medchemexpress, HY-N1579 1g Pyrogallol CAS:87-66-1 Pyrogallol is a polyphenol compound, which has anti-fungal and anti-psoriatic properties. Pyrogallol is a reductant that is able to generate free radicals, in particular superoxide anions. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3851