Learn More
Selleck Chemical LLC Pyrogallol 25mg 87-66-1 1,2,3-trihydroxybenzene
SDP
Catalog No. 501948429
Encompass_Preferred
Pyrogallol (1,2,3-trihydroxybenzene), an organic compound belonging to the phenol family, used as a photographic film developer and in the preparation of other chemicals. It is known to be a superoxide anion generator and ROS inducer. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8429 Supplier Selleck Chemical LLC Supplier No. S388525MG
Pyrogallol (1,2,3-trihydroxybenzene), an organic compound belonging to the phenol family, used as a photographic film developer and in the preparation of other chemicals. It is known to be a superoxide anion generator and ROS inducer. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.