Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pyrogallol

Catalog No. 25400210G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-400-210G 10 g
25-400-2250G 250 g
25-400-250G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 25-400-210G Supplier Sigma Aldrich Supplier No. 25400210G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Pyrogallol

Specifications

CAS 87-66-1
Melting Point 133°C to 134°C
Boiling Point 309°C
Molecular Formula C6H6O3
Linear Formula C6H3(OH)3
MDL Number MFCD00002192
Quantity 10 g
Synonym 1,2,3-Trihydroxybenzene; 2,3-Dihydroxyphenol; Pyrogallic acid
Molecular Weight (g/mol) 126.11 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.