Learn More
AdipoGen Pyrogallol
SDP
Catalog No. 502559894
Encompass_Preferred
Chemical. CAS 87-66-1. Formula C6H6O3. MW 126.11. The Polyphenol pyrogallol is a powerful naturally occuring reducing agent. Due to its oxygen radical generating property, pyrogallol is commonly used as a photographic developing agent, in hair dying industry, as an antiseptic and to calculate the amount of oxygen in the air. Pyrogallol is a potent superoxide anion O2•- generator. Pyrogallol causes oxidative damage and induces mutagenesis, carcinogenesis and hepatotoxicity. It impairs immune responses, inhibits phagocytosis and suppresses the proliferation due to its strong free radical generating potential. As a superoxide generator it is commonly used to investigate the role of free radicals in the biological system. It is a potent inducer of Nrf2-mediated gene expression. Pyrogallol possesses anti-bacterial, anti-fungal, anti-psoriatic anti-cancer, anti-inflammatory and neuroprotective properties.
Catalog No. 50-255-9894 Supplier AdipoGen Supplier No. CDXY0964G100
Chemical. CAS 87-66-1. Formula C6H6O3. MW 126.11. The Polyphenol pyrogallol is a powerful naturally occuring reducing agent. Due to its oxygen radical generating property, pyrogallol is commonly used as a photographic developing agent, in hair dying industry, as an antiseptic and to calculate the amount of oxygen in the air. Pyrogallol is a potent superoxide anion O2•- generator. Pyrogallol causes oxidative damage and induces mutagenesis, carcinogenesis and hepatotoxicity. It impairs immune responses, inhibits phagocytosis and suppresses the proliferation due to its strong free radical generating potential. As a superoxide generator it is commonly used to investigate the role of free radicals in the biological system. It is a potent inducer of Nrf2-mediated gene expression. Pyrogallol possesses anti-bacterial, anti-fungal, anti-psoriatic anti-cancer, anti-inflammatory and neuroprotective properties.