Learn More
Medchemexpress LLC Pyrogallol | 87-66-1 | MFCD00002192 | 99.6% | 126.11 g/mol | C6H6O3 | 10 MG
SDP
Catalog No. 5000295588
Encompass_Preferred
Pyrogallol (Standard) is a high-purity analytical reference standard of 1,2,3-benzenetriol supplied for research and analytical use. The compound is a polyphenolic reductant that can generate superoxide anions and is commonly used as a reference material in qualitative, quantitative, and methodological experiments in HPLC, GC, and MS.
  • High purity (99.6%) analytical standard.
  • Chemical identity: 1,2,3-benzenetriol; molecular weight 126.11 g/mol.
  • Suitable for HPLC, GC, and MS method development and calibration.
  • Supplied in accurately weighed small-quantity vials for reference use.
  • Storage and stability information provided in the certificate of analysis.
Catalog No. 50-002-95588 Supplier Medchemexpress LLC Supplier No. HYN1579R10MG
Pyrogallol (Standard) is a high-purity analytical reference standard of 1,2,3-benzenetriol supplied for research and analytical use. The compound is a polyphenolic reductant that can generate superoxide anions and is commonly used as a reference material in qualitative, quantitative, and methodological experiments in HPLC, GC, and MS.
  • High purity (99.6%) analytical standard.
  • Chemical identity: 1,2,3-benzenetriol; molecular weight 126.11 g/mol.
  • Suitable for HPLC, GC, and MS method development and calibration.
  • Supplied in accurately weighed small-quantity vials for reference use.
  • Storage and stability information provided in the certificate of analysis.