Learn More
Medchemexpress LLC Pyrogallol (Standard) | 87-66-1 | 126.11 | 25 MG
SDP
Catalog No. 5000389710
Encompass_Preferred
Pyrogallol (Standard) is the analytical standard of Pyrogallol, intended for research and analytical applications. It is a polyphenol compound with anti-fungal and anti-psoriatic properties. Pyrogallol acts as a reductant, capable of generating free radicals, specifically superoxide anions.
  • Analytical standard for research and analytical applications
  • Polyphenol compound with anti-fungal and anti-psoriatic properties
  • Acts as a reductant
  • Capable of generating free radicals, specifically superoxide anions
  • Used as a reference standard in assays
  • Commonly employed in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS
Catalog No. 50-003-89710 Supplier Medchemexpress LLC Supplier No. 25MGHYN1579R
Pyrogallol (Standard) is the analytical standard of Pyrogallol, intended for research and analytical applications. It is a polyphenol compound with anti-fungal and anti-psoriatic properties. Pyrogallol acts as a reductant, capable of generating free radicals, specifically superoxide anions.
  • Analytical standard for research and analytical applications
  • Polyphenol compound with anti-fungal and anti-psoriatic properties
  • Acts as a reductant
  • Capable of generating free radicals, specifically superoxide anions
  • Used as a reference standard in assays
  • Commonly employed in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS