Learn More
Lonza Walkersville PYROGENT Plus Bulk Gel Clot LAL Assay (with endotoxin)
Supplier:  Lonza Walkersville N494125
Encompass
The PYROGENT Gel Clot LAL Assay is a qualitative endotoxin detection test that gives a positive/negative result. If endotoxin is present, a gel clot will form and stay intact in a test tube when inverted 180°.Benefits:Sensitivity of 0.125 EU/ml Easy-to-read qualitative results No need to purchase CSE separatelyApplications:Simple LAL test not requiring sophisticated instrumentation and softwareContent:80 x 50 tests/vial lysate
Catalog No. NC0918151
Only null left
Add to Cart
Request a Quote