Learn More
Lonza Walkersville PYROGENT Plus Gel Clot LAL Assay (with endotoxin)
Supplier:  Lonza Walkersville N28306
Encompass
The PYROGENT Plus Gel Clot LAL Assay combines PYROGENT LAL with a matched control standard endotoxin together in one box kit. Standard kit sizes include 80 tests or 64 tests. The 80 and 64-test kits require depyrogentated 10 x 75 mm glass reaction tubes to run the assay. Benefits:Sensitivity of 0.06 EU/ml Easy-to-read qualitative results No need to purchase CSE separatelyApplications:Simple LAL test not requiring sophisticated instrumentation and softwareContent:4 x 16 tests/vial lysate
Catalog No. NC9711098