Learn More
Lonza Walkersville PYROGENT Plus Gel Clot LAL Assay (with endotoxin)
Catalog No. NC9758389
Encompass
The PYROGENT Plus Gel Clot LAL Assay combines PYROGENT LAL with a matched control standard endotoxin together in one box kit. Standard kit sizes include 4,000 tests, 250 tests, or 200 tests. The 4,000-, 250, and 200-test kits require depyrogentated 10 x 75 mm glass reaction tubes to run the assay.Benefits:Sensitivity of 0.03 EU/ml Easy-to-read qualitative results No need to purchase CSE separatelyApplications:Simple LAL test not requiring sophisticated instrumentation and softwareContent:4 x 50 tests/vial lysate
Catalog No. NC9758389 Supplier Lonza Walkersville Supplier No. N29403
The PYROGENT Plus Gel Clot LAL Assay combines PYROGENT LAL with a matched control standard endotoxin together in one box kit. Standard kit sizes include 4,000 tests, 250 tests, or 200 tests. The 4,000-, 250, and 200-test kits require depyrogentated 10 x 75 mm glass reaction tubes to run the assay.Benefits:Sensitivity of 0.03 EU/ml Easy-to-read qualitative results No need to purchase CSE separatelyApplications:Simple LAL test not requiring sophisticated instrumentation and softwareContent:4 x 50 tests/vial lysate