Learn More
Bioneer PYROHOTSTART TAQ PCR PREMIX, 0
SDP
Catalog No. NC2116354
Encompass
PyroHotStart Taq PCR PreMix, 0.2 ml thin-wall 8-tube strip with attached cap, 20 µl x 480 reactions
This item is not returnable. View return policy
Catalog No. NC2116354 Supplier Bioneer Supplier No. K2612
PyroHotStart Taq PCR PreMix, 0.2 ml thin-wall 8-tube strip with attached cap, 20 µl x 480 reactions