Learn More
MSC PYROLON PLUS 2 COVERALL
Catalog No. 5000768771
Encompass
Pyrolon Plus 2 garments offer Lakeland's lowest level of secondary flame-resistant protection. These garments are designed to help maintain the integrity of primary FR workwear. They are breathable and effectively repel water, grease, oil, and other potentially flammable hazards. These coveralls are suitable for work environments where primary FR clothing could be contaminated by hazardous or non-hazardous substances.
  • Provides the lowest level of secondary flame-resistant protection
  • Material is breathable and repels water, grease, oil, and potentially flammable hazards
  • Passes the Secondary FR Garment standard ANSI/ISEA 203
  • Third-party certified through UL
  • Suitable for environments with hazardous or non-hazardous substances that might contaminate primary FR clothing
Catalog No. 50-007-68771 Supplier MSC Supplier No. 61793717
Pyrolon Plus 2 garments offer Lakeland's lowest level of secondary flame-resistant protection. These garments are designed to help maintain the integrity of primary FR workwear. They are breathable and effectively repel water, grease, oil, and other potentially flammable hazards. These coveralls are suitable for work environments where primary FR clothing could be contaminated by hazardous or non-hazardous substances.
  • Provides the lowest level of secondary flame-resistant protection
  • Material is breathable and repels water, grease, oil, and potentially flammable hazards
  • Passes the Secondary FR Garment standard ANSI/ISEA 203
  • Third-party certified through UL
  • Suitable for environments with hazardous or non-hazardous substances that might contaminate primary FR clothing