Learn More
Electron Microscopy Sciences Pyrolytic Graphite Graphite Boat, B1
SDP
Catalog No. 5031649
Encompass_Preferred
Boat dimensions: Thickness of pyrolytic graphite foil is 0.51 mm (0.020") Overall length is 32 mm (1.25") and width is 9.6 mm (0.375”). Trough Dimension: 12.7 mm (L) x 9.0 mm (W) x 3.2 mm (D) (0.5 x 0.355 x 0.03”)
Catalog No. 50-316-49 Supplier Electron Microscopy Sciences Supplier No. 73808B1
Add to Cart
Add to Cart
Boat dimensions: Thickness of pyrolytic graphite foil is 0.51 mm (0.020") Overall length is 32 mm (1.25") and width is 9.6 mm (0.375”). Trough Dimension: 12.7 mm (L) x 9.0 mm (W) x 3.2 mm (D) (0.5 x 0.355 x 0.03”)