Learn More
Electron Microscopy Sciences SEM Pyrolytic Graphite Planchet 12.7 x 3.2 mm, 3.2 mm thick
SDP
Catalog No. 5031468
Encompass_Preferred
For SEM. Very light weight, 5'9's pure, solid crystal with no granular components. Withstands cryogenic and temperatures in excess of 3000 degree C. Smooth surface polished to 1 micron glassy finish.
Diameter: 12.7
Thickness: 3.2 mm
Catalog No. 50-314-68 Supplier Electron Microscopy Sciences Supplier No. 7629132
For SEM. Very light weight, 5'9's pure, solid crystal with no granular components. Withstands cryogenic and temperatures in excess of 3000 degree C. Smooth surface polished to 1 micron glassy finish.
Diameter: 12.7
Thickness: 3.2 mm