Learn More
Electron Microscopy Sciences Pyrolytic Graphite Stripper Film, 5 µm thick, 25.4 mm x 25.4 mm, 1.0 mg /sq cm pressure
SDP
Catalog No. 5031647
Encompass_Preferred
Smooth pyrolytic graphite stripper film is used in radiation sources, nuclear particle high energy density research, and medical diagnostic imaging instruments Pyrolytic graphite film offers major customer benefits relative to alternative materials such as amorphous carbon film or polycrystalline graphite films. These result in higher strength than alternative materials and films last 5 times longer than alternative materials.
Size: 25.4 mm x 25.4 mm
Thickness: 5 µm
Pressure: 1.0 mg /sq cm
Catalog No. 50-316-47 Supplier Electron Microscopy Sciences Supplier No. 7604105
Smooth pyrolytic graphite stripper film is used in radiation sources, nuclear particle high energy density research, and medical diagnostic imaging instruments Pyrolytic graphite film offers major customer benefits relative to alternative materials such as amorphous carbon film or polycrystalline graphite films. These result in higher strength than alternative materials and films last 5 times longer than alternative materials.
Size: 25.4 mm x 25.4 mm
Thickness: 5 µm
Pressure: 1.0 mg /sq cm