Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pyromellitic dianhydride glycerol dimethacrylate

Catalog No. 902101100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-210-1100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-210-1100G Supplier Sigma Aldrich Supplier No. 902101100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Pyromellitic dianhydride glycerol dimethacrylate (PMGDM) is an adhesive monomer synthesized from pyromellitic dianhydride and glycerol dimethacrylate. As a mixture of the para- and meta isomers, PMGDM has several advantages over more traditional adhesive monomers. PMGDM can be applied in thicker layers, and additives and/or additional initiators to improve curing can be readily incorporated.

Specifications

CAS 148019-46-9
Molecular Formula C32H34O16
Linear Formula C32H34O16
Quantity 100 g
Synonym PMGDM
Molecular Weight (g/mol) 674.6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.