Learn More
Cayman Chemical Pyronaridine (phosphate), Purity: ≥98%, CAS: 76748-86-2, 50 mg
SDP
Catalog No. 502970428
Encompass_Preferred
An antimalarial agent; active against six P. falciparum drug-sensitive or -resistant strains (IC50s = 1.53-3.94 nM); reduces parasitemia in P. chabaudi-infected mice at 3, 6, and 8 mg/kg in combination with artesunate; inhibits SARS-CoV-2 replication in infected A459-ACE2 cells (IC50 = 198 nM); cytotoxic against a panel of human cancer cells lines (CC50s = 1.6-4.9 µM)
Catalog No. 50-297-0428 Supplier Cayman Chemical Supplier No. 1613950 MG
An antimalarial agent; active against six P. falciparum drug-sensitive or -resistant strains (IC50s = 1.53-3.94 nM); reduces parasitemia in P. chabaudi-infected mice at 3, 6, and 8 mg/kg in combination with artesunate; inhibits SARS-CoV-2 replication in infected A459-ACE2 cells (IC50 = 198 nM); cytotoxic against a panel of human cancer cells lines (CC50s = 1.6-4.9 µM)