Learn More
Medchemexpress LLC HY-D0971 100mg Medchemexpress, Pyronin Y CAS:92-32-0 Purity:>98%
SDP
Catalog No. 501933218
Encompass_Preferred
Medchemexpress, HY-D0971 100mg Pyronin Y CAS:92-32-0 Pyronin Y is an intercalating cationic dye that shows specificity towards RNA. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3218 Supplier Medchemexpress LLC Supplier No. HYD0971100MG
Medchemexpress, HY-D0971 100mg Pyronin Y CAS:92-32-0 Pyronin Y is an intercalating cationic dye that shows specificity towards RNA. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.