Learn More
Biotang Inc PYRONIN Y (10MG/ML) 10 ML, BP-113, 10mg/ml Pyronin Y solution prepared in 18.2 megohms water and filtered through 0.22 micron filter.
Catalog No. 50152121 Shop All BIOTANG Inc Products
Encompass_Preferred
PYRONIN Y (10MG/ML) 10 ML, BP-113, 10mg/ml Pyronin Y solution prepared in 18.2 megohms water and filtered through 0.22 micron filter.
Catalog No. 50-152-121 Supplier Biotang Inc Supplier No. BTBB602
PYRONIN Y (10MG/ML) 10 ML, BP-113, 10mg/ml Pyronin Y solution prepared in 18.2 megohms water and filtered through 0.22 micron filter.