Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Pyronin Y, Certified, C.I. DcPy-2 5 GR
SDP

Catalog No. 5028210
Encompass_Preferred

(3-(N-Morpholino) propanesulfonic acid)
CAS #92-32-0 C₁₇H₁₉N₂O·Cl F.W. 302.8
Solubility: 9.0% Water; 0.5% Alcohol; 1.85% Cellosolve; 4.35% Glycol; 0.0% Xylene

Pyronin G. Cudder's method combined Gram/Pappenheim stain for gonorrheal pus. Kurnick's method used with methyl green which stains liver cells. For staining protein in the diazosulfanilic acid technique; substitute for azur A. Can be used for pyronin B.

Catalog No. 50-282-10 Supplier Electron Microscopy Sciences Supplier No. 19560
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(3-(N-Morpholino) propanesulfonic acid)
CAS #92-32-0 C₁₇H₁₉N₂O·Cl F.W. 302.8
Solubility: 9.0% Water; 0.5% Alcohol; 1.85% Cellosolve; 4.35% Glycol; 0.0% Xylene

Pyronin G. Cudder's method combined Gram/Pappenheim stain for gonorrheal pus. Kurnick's method used with methyl green which stains liver cells. For staining protein in the diazosulfanilic acid technique; substitute for azur A. Can be used for pyronin B.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.