Learn More
Weber Scientific PYRONIN Y-METHYL GRN STN,500ML
SDP
Catalog No. NC1876044
Encompass
Pyronin Y-Methyl Green Stain, 500 mlRequired for use when analyzing goat’s milk.
Catalog No. NC1876044 Supplier Weber Scientific Supplier No. 200260
Pyronin Y-Methyl Green Stain, 500 mlRequired for use when analyzing goat’s milk.