Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Pyronin Y, pure, high purity biological stain

Catalog No. AC200100050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC200100050 5 g Glass bottle
AC200100010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC200100050 Supplier Thermo Scientific Chemicals Supplier No. 200100050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 92-32-0 | C17H19ClN2O | 302.802 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-32-0
Molecular Formula C17H19ClN2O
Molecular Weight (g/mol) 302.802
MDL Number MFCD00011725
InChI Key INCIMLINXXICKS-UHFFFAOYSA-M
Synonym C.I. 45005, Pyronin G
PubChem CID 7085
ChEBI CHEBI:87347
IUPAC Name [6-(dimethylamino)xanthen-3-ylidene]-dimethylazanium;chloride
SMILES CN(C)C1=CC2=C(C=C1)C=C3C=CC(=[N+](C)C)C=C3O2.[Cl-]

Specifications

Absorption Ratio 1.23 to 1.7
Dye Content 50% min.
Infrared Spectrum Authentic
Lambda Max 546 to 551nm (in 50% ethanol)
Packaging Glass bottle
Beilstein 18, V,10, 181
Merck Index 15, 8119
Color Green
Quantity 5 g
Physical Form Crystalline Powder
Formula Weight 302.79
Grade Pure
Chemical Name or Material Pyronin Y, Pure
Show More Show Less
Hazard Statement GHS H Statement:
Suspected of causing genetic defects.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.