Learn More
Abcam Pyronin Y (Pyronin G), Cationic dye (RNA), 5G
Catalog No. 502704692
Encompass_Preferred
MW 302.8 g/mol. Cationic dye that intercalates RNA and is visible as a red band during electrophoresis. Shown to accumulate in the mitochondria of viable cells. At low concentrations the cytostatic effect is cell arrest in the G1 phase, whereas at high concentrations the cytotoxic effect is cell arrest in the G2 and S phase. Used in combination with Alcian Blue to assess and study pulmonary diseases, and in combination with Methyl Green to stain nucleic acids in paraffin sections. Additionally, has been used to measure RNA in CHO cells by flow cytometry.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4692 Supplier Abcam Supplier No. AB1463505G
MW 302.8 g/mol. Cationic dye that intercalates RNA and is visible as a red band during electrophoresis. Shown to accumulate in the mitochondria of viable cells. At low concentrations the cytostatic effect is cell arrest in the G1 phase, whereas at high concentrations the cytotoxic effect is cell arrest in the G2 and S phase. Used in combination with Alcian Blue to assess and study pulmonary diseases, and in combination with Methyl Green to stain nucleic acids in paraffin sections. Additionally, has been used to measure RNA in CHO cells by flow cytometry.
The product is subject to the following: Abcam Restricted Use Statement