Learn More
Medchemexpress LLC Pyronine B | 500 MG
SDP
Catalog No. 5000409810
Encompass_Preferred
Pyronine B is an organic cationic dye that can be used as a solution-processable and heating-free n-type dopant for soft electronics.
  • Organic cationic dye
  • Solution-processable n-type dopant
  • Heating-free n-type dopant
  • Used for soft electronics
  • Fluorescent dye
Catalog No. 50-004-09810 Supplier Medchemexpress LLC Supplier No. HYD0949500MG
Pyronine B is an organic cationic dye that can be used as a solution-processable and heating-free n-type dopant for soft electronics.
  • Organic cationic dye
  • Solution-processable n-type dopant
  • Heating-free n-type dopant
  • Used for soft electronics
  • Fluorescent dye