Learn More
Medchemexpress LLC Pyropheophorbide-a | 24533-72-0 | 98.0% | 1 ML
SDP
Catalog No. 5000340155
Encompass_Preferred
Pyropheophorbide-a (Ppa) is a potent photosensitizer primarily used in photodynamic therapy (PDT) for the treatment of various tumors. This compound demonstrates significant phototoxic effects on tumor cells, such as cervical cancer cells, and also exhibits anti-lipogenesis activity. Its potential applications extend to research in cancer and age-related macular degeneration (AMD), with derivatives also showing inhibitory activity against tumor cells.
  • Photosensitizer for photodynamic therapy
  • Exhibits phototoxic effects on tumor cells
  • Demonstrates anti-lipogenesis activity
  • Potential for research in cancer and age-related macular degeneration (AMD)
  • Derivatives show inhibitory activity against tumor cells
Catalog No. 50-003-40155 Supplier Medchemexpress LLC Supplier No. HY12897310MM/1ML
Pyropheophorbide-a (Ppa) is a potent photosensitizer primarily used in photodynamic therapy (PDT) for the treatment of various tumors. This compound demonstrates significant phototoxic effects on tumor cells, such as cervical cancer cells, and also exhibits anti-lipogenesis activity. Its potential applications extend to research in cancer and age-related macular degeneration (AMD), with derivatives also showing inhibitory activity against tumor cells.
  • Photosensitizer for photodynamic therapy
  • Exhibits phototoxic effects on tumor cells
  • Demonstrates anti-lipogenesis activity
  • Potential for research in cancer and age-related macular degeneration (AMD)
  • Derivatives show inhibitory activity against tumor cells