Learn More
Cayman Chemical PyropheophorbIde a methyl25mg
SDP
Supplier:  Cayman Chemical 3651225 MG
Encompass_Preferred
A photosensitizer; induces cell death in RIF-1 cells when used in combination with PDT at 0.06 and 1 µM; induces IL-1 receptor internalization and NF-κB activation following light exposure in HCT116 cells
Catalog No. 50-291-1580