Learn More
Cayman Chemical Pyropheophorbide a methyl ester, Purity: ≥95%, CAS: 6453-67-4, 5 mg
SDP
Catalog No. 502911840
Encompass_Preferred
A photosensitizer; induces cell death in RIF-1 cells when used in combination with PDT at 0.06 and 1 µM; induces IL-1 receptor internalization and NF-κB activation following light exposure in HCT116 cells
Catalog No. 50-291-1840 Supplier Cayman Chemical Supplier No. 365125 MG
A photosensitizer; induces cell death in RIF-1 cells when used in combination with PDT at 0.06 and 1 µM; induces IL-1 receptor internalization and NF-κB activation following light exposure in HCT116 cells