Learn More
New England Biolabs, Inc. Pyrophosphatase, Inorganic (E. coli) – 50 units
SDP
Catalog No. 504384
Encompass
Inorganic pyrophosphatase (PPase) catalyzes the hydrolysis of inorganic pyrophosphate to form orthophosphate:
Catalog No. 50-438-4 Supplier New England Biolabs, Inc. Supplier No. M0361L
 Restricted Item
Add to Cart
Add to Cart
Inorganic pyrophosphatase (PPase) catalyzes the hydrolysis of inorganic pyrophosphate to form orthophosphate: