Learn More
New England Biolabs, Inc. Pyrophosphatase, inorganic (yeast) – 50 units
SDP
Catalog No. 50995251
Encompass
Inorganic pyrophosphatase (PPase) catalyzes the hydrolysis of inorganic pyrophosphate to form orthophosphate. A variety of metabolic reactions generate inorganic pyrophosphate as a reaction byproduct. Such reactions are rendered irreversible when the pyrophosphate is degraded by pyrophosphatase. RNA and DNA synthesis are examples of reactions that can be pulled far in the synthesis direction by the action of inorganic pyrophosphatase.
Catalog No. 50-995-251 Supplier New England Biolabs, Inc. Supplier No. M2403L
Inorganic pyrophosphatase (PPase) catalyzes the hydrolysis of inorganic pyrophosphate to form orthophosphate. A variety of metabolic reactions generate inorganic pyrophosphate as a reaction byproduct. Such reactions are rendered irreversible when the pyrophosphate is degraded by pyrophosphatase. RNA and DNA synthesis are examples of reactions that can be pulled far in the synthesis direction by the action of inorganic pyrophosphatase.