Learn More
ABclonal Technology Pyrophosphatase 1 Rabbit mAb
SDP
Catalog No. 5000008008
Encompass_Preferred
The protein encoded by this gene is a member of the inorganic pyrophosphatase (PPase) family PPases catalyze the hydrolysis of pyrophosphate to inorganic phosphate which is important for the phosphate metabolism of cells Studies of a similar protein in bovine suggested a cytoplasmic localization of this enzyme
Catalog No. 50-000-08008 Supplier ABclonal Technology Supplier No. A20942100UL
The protein encoded by this gene is a member of the inorganic pyrophosphatase (PPase) family PPases catalyze the hydrolysis of pyrophosphate to inorganic phosphate which is important for the phosphate metabolism of cells Studies of a similar protein in bovine suggested a cytoplasmic localization of this enzyme