Learn More
Cell Signaling Technology Pyroptosis Antibody Sampler Kit, 1 Kit
SDP
Catalog No. 502051346
Encompass
The Pyroptosis Antibody Sampler Kit provides an economical solution for detecting key proteins used as readouts for pyroptosis. This kit includes a comprehensive selection of primary antibodies to facilitate the study of this regulated inflammatory cell death pathway via western blotting.
  • Includes enough material to perform two western blot experiments per primary antibody
  • Features antibodies for pyroptosis markers including gasdermin D, caspase-1, IL-1β, and HMGB1
  • Supplied with HRP-linked secondary antibody for convenient detection
  • Supports analysis of canonical and non-canonical inflammasome components
Catalog No. 50-205-1346 Supplier Cell Signaling Technology Supplier No. 43811T
The Pyroptosis Antibody Sampler Kit provides an economical solution for detecting key proteins used as readouts for pyroptosis. This kit includes a comprehensive selection of primary antibodies to facilitate the study of this regulated inflammatory cell death pathway via western blotting.
  • Includes enough material to perform two western blot experiments per primary antibody
  • Features antibodies for pyroptosis markers including gasdermin D, caspase-1, IL-1β, and HMGB1
  • Supplied with HRP-linked secondary antibody for convenient detection
  • Supports analysis of canonical and non-canonical inflammasome components