Learn More
Associates of Cape Cod Inc Pyrosate Kit, 0.03 EU/mL 30-Test Bulk Package
SDP
Catalog No. NC1241127
Encompass
Includes sample test tubes and positive product control test tubes. The Pyrosate kit is developed as an easy-to-use LAL gel-clot test. The assay does not require special training or laboratory supplies. The step-by-step illustrated instructions allow the user to perform assays within minutes. The Pyrosate kit provides rapid results and is especially convenient for research, testing water and dialysate.
Catalog No. NC1241127 Supplier Associates of Cape Cod Inc Supplier No. PSD03030
Includes sample test tubes and positive product control test tubes. The Pyrosate kit is developed as an easy-to-use LAL gel-clot test. The assay does not require special training or laboratory supplies. The step-by-step illustrated instructions allow the user to perform assays within minutes. The Pyrosate kit provides rapid results and is especially convenient for research, testing water and dialysate.