Learn More
Associates of Cape Cod Inc Pyrosol Buffer with pH indicator (gel-clot method only), 5.5 mL/vial 5 Pack
SDP
Catalog No. NC0268423
Encompass
Pyrosol is an FDA licensed buffer for reconstituting Pyrotell multi-test or Pyrotell-T lysate. It is used for testing electrolytes, strongly buffered solutions (especially bicarbonate buffers), and solutions for which it is difficult to adjust pH into the required range. Pyrosol buffer is also available with a pH indicator for gel-clot applications.
Catalog No. NC0268423 Supplier Associates of Cape Cod Inc Supplier No. BC0515
Pyrosol is an FDA licensed buffer for reconstituting Pyrotell multi-test or Pyrotell-T lysate. It is used for testing electrolytes, strongly buffered solutions (especially bicarbonate buffers), and solutions for which it is difficult to adjust pH into the required range. Pyrosol buffer is also available with a pH indicator for gel-clot applications.