Learn More
Associates of Cape Cod Inc PYROSOL BUFFER W/PH 5.5ML 5PK
SDP
Catalog No. NC0253714
Encompass
PYROSOL RECONSTITUTION BUFFER WITH pH INDICATOR, FOR USE WITH GEL CLOT ONLY, 5.5ML/VIAL, 5 VIALS/PACK, PYROCLEAR CERTIFIED
Catalog No. NC0253714 Supplier Associates of Cape Cod Inc Supplier No. BR0515
PYROSOL RECONSTITUTION BUFFER WITH pH INDICATOR, FOR USE WITH GEL CLOT ONLY, 5.5ML/VIAL, 5 VIALS/PACK, PYROCLEAR CERTIFIED