Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Associates of Cape Cod Inc Pyrosol LAL Reconstitution Buffer, 5.5 mL per vial, 25 vials, pH indicator included
SDP

Catalog No. NC0851762
Encompass

The Pyrosol LAL Reconstitution Buffer is a specialized FDA-licensed buffer from Associates of Cape Cod, Inc., designed for precise reconstitution of Pyrotell multi-test or Pyrotell-T lysate. It facilitates accurate endotoxin testing for samples that may pose challenges in pH adjustment, such as electrolytes and strongly buffered solutions. The included pH indicator ensures reliable gel-clot applications, enhancing testing accuracy and consistency.

  • Supplier: Associates of Cape Cod, Inc.
  • Product type: LAL reconstitution buffer
  • Application: Gel-clot method only
  • Volume per vial: 5.5 mL
  • Pack size: 25 vials
  • pH indicator: Included
  • Licensing: FDA licensed
  • Usage: Reconstitutes Pyrotell multi-test or Pyrotell-T lysate
  • Compatibility: Electrolytes, strongly buffered solutions
  • Benefit: Enhances accuracy and consistency in endotoxin testing
  • Feature: Provides extra pH buffering capacity

Catalog No. NC0851762 Supplier Associates of Cape Cod Inc Supplier No. BR05125
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Pyrosol LAL Reconstitution Buffer is a specialized FDA-licensed buffer from Associates of Cape Cod, Inc., designed for precise reconstitution of Pyrotell multi-test or Pyrotell-T lysate. It facilitates accurate endotoxin testing for samples that may pose challenges in pH adjustment, such as electrolytes and strongly buffered solutions. The included pH indicator ensures reliable gel-clot applications, enhancing testing accuracy and consistency.

  • Supplier: Associates of Cape Cod, Inc.
  • Product type: LAL reconstitution buffer
  • Application: Gel-clot method only
  • Volume per vial: 5.5 mL
  • Pack size: 25 vials
  • pH indicator: Included
  • Licensing: FDA licensed
  • Usage: Reconstitutes Pyrotell multi-test or Pyrotell-T lysate
  • Compatibility: Electrolytes, strongly buffered solutions
  • Benefit: Enhances accuracy and consistency in endotoxin testing
  • Feature: Provides extra pH buffering capacity

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.